siRNA Details
Gene Symbol: Gpr37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057201.1 | NP_476549.1 | 1 | 1505 | ||
GenBank mRNA | AF087946.1 | - | 1 | 1505 | ||
BC087728.1 | - | 1 | 1571 | 7320414 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
