siRNA Details
| Gene Symbol: Kif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057202.1 | NP_476550.1 | 19 | 2116 | ||
| XM_006254005.1 | XP_006254067.1 | 19 | 2547 | |||
| GenBank mRNA | AF155822.1 | - | 5 | 467 | ||
| DQ309275.1 | - | 19 | 2116 | |||

Kif5b
Genomic Map