siRNA Details
Gene Symbol: Pnliprp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057206.1 | NP_476554.1 | 9, 10 | 1172 | ||
GenBank mRNA | L09216.1 | - | 9, 10 | 1172 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pnliprp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057206.1 | NP_476554.1 | 9, 10 | 1172 | ||
GenBank mRNA | L09216.1 | - | 9, 10 | 1172 |