siRNA Details
| Gene Symbol: Sv2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057207.3 | NP_476555.1 | 6 | 1408 | ||
| XM_008759492.1 | XP_008757714.1 | 6 | 2127 | |||
| XM_008759493.1 | XP_008757715.1 | 6 | 2505 | |||
| XM_008759494.1 | XP_008757716.1 | 6 | 3027 | |||
| XM_008759495.1 | XP_008757717.1 | 6 | 2123 | |||
| GenBank mRNA | L10362.1 | - | 6 | 1408 | ||

Sv2b
Genomic Map