siRNA Details
| Gene Symbol: Mylk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057209.1 | NP_476557.1 | 2, 3 | 578 | ||
| GenBank mRNA | J03886.1 | - | 2, 3 | 578 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mylk2| Gene Symbol: Mylk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057209.1 | NP_476557.1 | 2, 3 | 578 | ||
| GenBank mRNA | J03886.1 | - | 2, 3 | 578 | ||