siRNA Details
| Gene Symbol: Lama5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191609.1 | NP_001178538.1 | 60 | 8085 | ||
| GenBank mRNA | FQ223406.1 | - | 6 | 610 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lama5| Gene Symbol: Lama5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191609.1 | NP_001178538.1 | 60 | 8085 | ||
| GenBank mRNA | FQ223406.1 | - | 6 | 610 | ||