siRNA Details
| Gene Symbol: Chek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080400.1 | NP_536325.1 | 13 | 1458 | ||
| XM_008766042.1 | XP_008764264.1 | 13 | 2591 | |||
| XM_008766043.1 | XP_008764265.1 | 13 | 1528 | |||
| GenBank mRNA | AF414135.1 | - | 13 | 1458 | ||
| AF414136.2 | - | 8 | 877 | |||
| AF443592.1 | - | 13 | 1456 | |||

Chek1
Genomic Map