siRNA Details
| Gene Symbol: Gli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191910.1 | NP_001178839.1 | 3 | 225 | ||
| XM_006241442.2 | XP_006241504.1 | 5 | 490 | |||
| XM_006241443.2 | XP_006241505.1 | 4 | 373 | |||
| XM_008765375.1 | XP_008763597.1 | 5 | 438 | |||

Gli1
Genomic Map