siRNA Details
| Gene Symbol: Sytl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080410.1 | NP_536335.1 | 3 | 371 | ||
| XM_006257213.2 | XP_006257275.1 | 4 | 579 | |||
| XM_008773387.1 | XP_008771609.1 | 4 | 579 | |||
| GenBank mRNA | AF419341.1 | - | 3 | 371 | ||
| AF419342.1 | - | 3 | 371 | |||

Sytl4
Genomic Map