siRNA Details
| Gene Symbol: Nploc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080577.2 | NP_542144.1 | 12 | 1350 | ||
| GenBank mRNA | AF234600.1 | - | 12 | 1278 | ||
| BC101887.1 | - | 12 | 1350 | 7930738 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nploc4| Gene Symbol: Nploc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080577.2 | NP_542144.1 | 12 | 1350 | ||
| GenBank mRNA | AF234600.1 | - | 12 | 1278 | ||
| BC101887.1 | - | 12 | 1350 | 7930738 | ||