siRNA Details
Gene Symbol: Ap3m2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133305.2 | NP_579839.1 | 5 | 864 | ||
GenBank mRNA | BC086993.1 | - | 5 | 864 | 7131933 | |
L07074.1 | - | 687 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ap3m2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133305.2 | NP_579839.1 | 5 | 864 | ||
GenBank mRNA | BC086993.1 | - | 5 | 864 | 7131933 | |
L07074.1 | - | 687 |