siRNA Details
| Gene Symbol: Gabrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080586.1 | NP_542153.1 | 9 | 1249 | ||
| GenBank mRNA | X57514.1 | - | 9 | 1249 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrg1| Gene Symbol: Gabrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080586.1 | NP_542153.1 | 9 | 1249 | ||
| GenBank mRNA | X57514.1 | - | 9 | 1249 | ||