siRNA Details
| Gene Symbol: Olr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133306.2 | NP_579840.2 | 4 | 633 | ||
| GenBank mRNA | AB005900.1 | - | 4 | 633 | ||
| BC097290.1 | - | 4 | 586 | 7458637 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1| Gene Symbol: Olr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133306.2 | NP_579840.2 | 4 | 633 | ||
| GenBank mRNA | AB005900.1 | - | 4 | 633 | ||
| BC097290.1 | - | 4 | 586 | 7458637 | ||