siRNA Details
| Gene Symbol: Slco3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177481.1 | NP_803434.1 | 8 | 1563 | ||
| XM_006229378.2 | XP_006229440.1 | 8 | 1728 | |||
| GenBank mRNA | AF239219.1 | - | 8 | 1563 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco3a1| Gene Symbol: Slco3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177481.1 | NP_803434.1 | 8 | 1563 | ||
| XM_006229378.2 | XP_006229440.1 | 8 | 1728 | |||
| GenBank mRNA | AF239219.1 | - | 8 | 1563 | ||