siRNA Details
| Gene Symbol: Bnip3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080888.1 | NP_543164.1 | 4, 5 | 462 | ||
| GenBank mRNA | AB077364.1 | - | 4, 5 | 462 | ||
| AF441118.1 | - | 4, 5 | 531 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bnip3l| Gene Symbol: Bnip3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080888.1 | NP_543164.1 | 4, 5 | 462 | ||
| GenBank mRNA | AB077364.1 | - | 4, 5 | 462 | ||
| AF441118.1 | - | 4, 5 | 531 | |||