siRNA Details
| Gene Symbol: Daxx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080891.2 | NP_543167.2 | 3 | 552 | ||
| XM_006256103.2 | XP_006256165.1 | 3 | 1334 | |||
| XM_006256104.2 | XP_006256166.1 | 3 | 1008 | |||
| XM_006256105.2 | XP_006256167.1 | 3 | 904 | |||
| XM_006256106.2 | XP_006256168.1 | 3 | 570 | |||
| XM_006256107.2 | XP_006256169.1 | 3 | 674 | |||
| XM_006256108.2 | XP_006256170.1 | 3 | 554 | |||
| XM_006256109.1 | XP_006256171.1 | 2 | 1427 | |||
| XM_006256110.2 | XP_006256172.1 | 3 | 553 | |||
| XM_008772704.1 | XP_008770926.1 | 2 | 1427 | |||
| GenBank mRNA | AB064671.1 | - | 2 | 456 | ||
| BC128729.1 | - | 3 | 552 | 8366304 | ||
| FQ223571.1 | - | 3 | 523 | |||

Daxx
Genomic Map