siRNA Details
Gene Symbol: Pde7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080894.2 | NP_543170.1 | 12, 13 | 1215 | ||
XM_008758591.1 | XP_008756813.1 | 11, 12 | 1535 | |||
XM_008758592.1 | XP_008756814.1 | 11, 12 | 1119 | |||
GenBank mRNA | AB057409.1 | - | 12, 13 | 1215 | ||
AB057410.1 | - | 11, 12 | 1083 | |||
AB057411.1 | - | 13, 14 | 1184 |
