siRNA Details
| Gene Symbol: Afap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080900.1 | NP_543176.1 | 11 | 1505 | ||
| XM_008770245.1 | XP_008768467.1 | 12 | 1664 | |||
| XM_008770246.1 | XP_008768468.1 | 12 | 1664 | |||
| XM_008770247.1 | XP_008768469.1 | 12 | 1667 | |||
| GenBank mRNA | AY063759.1 | - | 11 | 1505 | ||

Afap1
Genomic Map