siRNA Details
| Gene Symbol: Afap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080900.1 | NP_543176.1 | 9 | 1211 | ||
| XM_008770245.1 | XP_008768467.1 | 10 | 1370 | |||
| XM_008770246.1 | XP_008768468.1 | 10 | 1370 | |||
| XM_008770247.1 | XP_008768469.1 | 10 | 1373 | |||
| GenBank mRNA | AY063759.1 | - | 9 | 1211 | ||

Afap1
Genomic Map