siRNA Details
Gene Symbol: Higd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080902.4 | NP_543178.2 | 2 | 137 | ||
GenBank mRNA | AY062253.3 | - | 2 | 196 | ||
BC059118.1 | - | 2 | 141 | 6890532 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Higd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080902.4 | NP_543178.2 | 2 | 137 | ||
GenBank mRNA | AY062253.3 | - | 2 | 196 | ||
BC059118.1 | - | 2 | 141 | 6890532 |