siRNA Details
| Gene Symbol: Siah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080905.2 | NP_543181.2 | 2 | 193 | ||
| XM_006255211.2 | XP_006255273.1 | 2 | 4794 | |||
| XM_006255213.2 | XP_006255275.1 | 2 | 533 | |||
| XM_008772345.1 | XP_008770567.1 | 3 | 234 | |||
| GenBank mRNA | AB067814.1 | - | 2 | 52 | ||
| AY681965.1 | - | 1 | 13 | |||
| DV213782.1 | - | 2 | 193 | |||

Siah1
Genomic Map