siRNA Details
| Gene Symbol: Cyp26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130408.2 | NP_569092.2 | 7 | 1435 | ||
| GenBank mRNA | AF439720.2 | - | 7 | 1435 | ||
| DQ266888.1 | - | 7 | 1505 | |||
| DQ317305.1 | - | 7 | 1445 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp26a1| Gene Symbol: Cyp26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130408.2 | NP_569092.2 | 7 | 1435 | ||
| GenBank mRNA | AF439720.2 | - | 7 | 1435 | ||
| DQ266888.1 | - | 7 | 1505 | |||
| DQ317305.1 | - | 7 | 1445 | |||