siRNA Details
| Gene Symbol: Coro1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130411.2 | NP_569095.1 | 2, 3 | 305 | ||
| XM_006230204.2 | XP_006230266.1 | 2, 3 | 389 | |||
| GenBank mRNA | AF416730.1 | - | 1, 2 | 201 | ||
| AF495469.1 | - | 2, 3 | 305 | |||
| BC086971.1 | - | 2, 3 | 281 | 7124429 | ||
| FQ226379.1 | - | 2, 3 | 293 | |||
| FQ226941.1 | - | 2, 3 | 325 | |||
| FQ233306.1 | - | 2, 3 | 299 | |||
| FQ233390.1 | - | 2, 3 | 301 | |||
| FQ233414.1 | - | 2, 3 | 303 | |||
| FQ234750.1 | - | 2, 3 | 303 | |||

Coro1a
Genomic Map