siRNA Details
Gene Symbol: Skap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130413.1 | NP_569097.1 | 1, 2 | 199 | ||
XM_006236477.2 | XP_006236539.1 | 1, 2 | 184 | |||
XM_006236478.2 | XP_006236540.1 | 1, 2 | 184 | |||
GenBank mRNA | AF302132.1 | - | 1, 2 | 199 | ||
BC070949.1 | - | 1, 2 | 146 | 7099436 |
