siRNA Details
Gene Symbol: Dcbld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130419.1 | NP_569103.1 | 10 | 1275 | ||
GenBank mRNA | AF387549.1 | - | 10 | 1275 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dcbld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130419.1 | NP_569103.1 | 10 | 1275 | ||
GenBank mRNA | AF387549.1 | - | 10 | 1275 |