siRNA Details
Gene Symbol: Lcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130421.2 | NP_569105.1 | 10, 11 | 908 | ||
GenBank mRNA | AB072980.1 | - | 10, 11 | 748 | ||
BC100066.1 | - | 10, 11 | 908 | 7387128 | ||
FQ232596.1 | - | 10, 11 | 922 |
