siRNA Details
Gene Symbol: Acaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130433.1 | NP_569117.1 | 3, 4 | 467 | ||
GenBank mRNA | FQ216726.1 | - | 3, 4 | 330 | ||
FQ228855.1 | - | 3, 4 | 332 | |||
X05341.1 | - | 3, 4 | 467 |
