siRNA Details
| Gene Symbol: Galnt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130742.1 | NP_570098.1 | 7 | 972 | ||
| GenBank mRNA | AF241241.1 | - | 7 | 972 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galnt10| Gene Symbol: Galnt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130742.1 | NP_570098.1 | 7 | 972 | ||
| GenBank mRNA | AF241241.1 | - | 7 | 972 | ||