siRNA Details
| Gene Symbol: Ryr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762132.1 | XP_008760354.1 | 49 | 7484 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ryr3| Gene Symbol: Ryr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762132.1 | XP_008760354.1 | 49 | 7484 | ||