siRNA Details
| Gene Symbol: Fgf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130814.1 | NP_570827.1 | 5 | 713 | ||
| GenBank mRNA | AB079374.1 | - | 5 | 713 | ||
| AF348446.1 | - | 4 | 527 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf12| Gene Symbol: Fgf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130814.1 | NP_570827.1 | 5 | 713 | ||
| GenBank mRNA | AB079374.1 | - | 5 | 713 | ||
| AF348446.1 | - | 4 | 527 | |||