siRNA Details
Gene Symbol: Samsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130821.1 | NP_570834.1 | 4 | 347 | ||
GenBank mRNA | AB067679.1 | - | 4 | 347 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Samsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130821.1 | NP_570834.1 | 4 | 347 | ||
GenBank mRNA | AB067679.1 | - | 4 | 347 |