siRNA Details
| Gene Symbol: Nphs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130828.2 | NP_570841.2 | 6 | 841 | ||
| GenBank mRNA | AB091379.1 | - | 6 | 780 | ||
| AF309631.1 | - | 5 | 519 | |||
| AY039651.1 | - | 6 | 841 | |||
| BC098649.1 | - | 6 | 823 | 7127895 | ||

Nphs2
Genomic Map