siRNA Details
| Gene Symbol: Atp2c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131907.2 | NP_571982.2 | 25 | 2764 | ||
| GenBank mRNA | BC086994.1 | - | 25 | 2764 | 7106233 | |
| M93017.1 | - | 25 | 2475 | |||
| M93018.1 | - | 1891 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp2c1| Gene Symbol: Atp2c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131907.2 | NP_571982.2 | 25 | 2764 | ||
| GenBank mRNA | BC086994.1 | - | 25 | 2764 | 7106233 | |
| M93017.1 | - | 25 | 2475 | |||
| M93018.1 | - | 1891 | ||||