siRNA Details
Gene Symbol: Fgf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131908.1 | NP_571983.1 | 2 | 418 | ||
GenBank mRNA | AB079674.1 | - | 2 | 418 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fgf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131908.1 | NP_571983.1 | 2 | 418 | ||
GenBank mRNA | AB079674.1 | - | 2 | 418 |