siRNA Details
Gene Symbol: Anp32b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131911.2 | NP_571986.1 | 2 | 361 | ||
XM_008763695.1 | XP_008761917.1 | 3 | 673 | |||
GenBank mRNA | AB025581.1 | - | 2 | 198 | ||
BC086508.1 | - | 2 | 361 | 7103152 |
