siRNA Details
| Gene Symbol: Kcnh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131912.1 | NP_571987.1 | 2 | 404 | ||
| XM_006234269.2 | XP_006234331.1 | 2 | 469 | |||
| GenBank mRNA | AF016191.1 | - | 2 | 404 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnh7| Gene Symbol: Kcnh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131912.1 | NP_571987.1 | 2 | 404 | ||
| XM_006234269.2 | XP_006234331.1 | 2 | 469 | |||
| GenBank mRNA | AF016191.1 | - | 2 | 404 | ||