siRNA Details
| Gene Symbol: Capn8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133309.2 | NP_579843.2 | 15 | 1733 | ||
| GenBank mRNA | D14478.1 | - | 17 | 1963 | ||
| D14479.1 | - | 15 | 1733 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Capn8| Gene Symbol: Capn8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133309.2 | NP_579843.2 | 15 | 1733 | ||
| GenBank mRNA | D14478.1 | - | 17 | 1963 | ||
| D14479.1 | - | 15 | 1733 | |||