siRNA Details
| Gene Symbol: Olr59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173293.1 | NP_775415.1 | 2 | 658 | ||
| XM_008759669.1 | XP_008757891.1 | 3 | 2115 | |||
| XM_008759670.1 | XP_008757892.1 | 3 | 893 | |||
| XM_008759671.1 | XP_008757893.1 | 3 | 322 | |||
| GenBank mRNA | AF079864.1 | - | 2 | 658 | ||

Olr59
Genomic Map