siRNA Details
| Gene Symbol: Prdm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133312.1 | NP_579846.1 | 3 | 1015 | ||
| XM_006241160.2 | XP_006241222.1 | 5 | 1477 | |||
| XM_006241161.1 | XP_006241223.1 | 5 | 1476 | |||
| XM_008765213.1 | XP_008763435.1 | 5 | 1482 | |||
| GenBank mRNA | AF176023.1 | - | 3 | 1015 | ||

Prdm4
Genomic Map