siRNA Details
| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133313.2 | NP_579847.1 | 6, 7 | 830 | ||
| GenBank mRNA | AJ301634.1 | - | 6, 7 | 939 | ||
| BC062087.1 | - | 6, 7 | 830 | 5621521 | ||
| FQ213088.1 | - | 1, 2 | 62 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atxn10| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133313.2 | NP_579847.1 | 6, 7 | 830 | ||
| GenBank mRNA | AJ301634.1 | - | 6, 7 | 939 | ||
| BC062087.1 | - | 6, 7 | 830 | 5621521 | ||
| FQ213088.1 | - | 1, 2 | 62 | |||