siRNA Details
| Gene Symbol: Slc40a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133315.2 | NP_579849.2 | 3 | 530 | ||
| GenBank mRNA | AF394785.3 | - | 3 | 475 | ||
| CK364414.1 | - | 2 | 162 | |||
| FQ226522.1 | - | 2 | 190 | |||
| U76714.1 | - | 3 | 545 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc40a1| Gene Symbol: Slc40a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133315.2 | NP_579849.2 | 3 | 530 | ||
| GenBank mRNA | AF394785.3 | - | 3 | 475 | ||
| CK364414.1 | - | 2 | 162 | |||
| FQ226522.1 | - | 2 | 190 | |||
| U76714.1 | - | 3 | 545 | |||