siRNA Details
| Gene Symbol: Mutyh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133316.1 | NP_579850.1 | 13, 14 | 1290 | ||
| XM_006238626.2 | XP_006238688.1 | 13, 14 | 1318 | |||
| XM_006238627.2 | XP_006238689.1 | 12, 13 | 1215 | |||
| XM_008763987.1 | XP_008762209.1 | 11, 12 | 1276 | |||
| GenBank mRNA | AF478683.1 | - | 13, 14 | 1290 | ||
| BC128728.1 | - | 11, 12 | 1442 | |||

Mutyh
Genomic Map