siRNA Details
| Gene Symbol: Mutyh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133316.1 | NP_579850.1 | 2 | 70 | ||
| XM_006238626.2 | XP_006238688.1 | 2 | 98 | |||
| XM_006238627.2 | XP_006238689.1 | 2 | 109 | |||
| GenBank mRNA | AF478683.1 | - | 2 | 70 | ||
| BC128728.1 | - | 2 | 40 | |||
| FQ230684.1 | - | 2 | 106 | |||

Mutyh
Genomic Map