siRNA Details
| Gene Symbol: Zfp111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133323.1 | NP_579857.1 | 4 | 535 | ||
| XM_006228375.2 | XP_006228437.1 | 2 | 475 | |||
| XM_006228376.2 | XP_006228438.1 | 3 | 631 | |||
| GenBank mRNA | FQ211918.1 | - | 4 | 601 | ||
| U27186.1 | - | 4 | 535 | |||

Zfp111
Genomic Map