siRNA Details
Gene Symbol: Zfp111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133323.1 | NP_579857.1 | 4 | 591 | ||
GenBank mRNA | U27186.1 | - | 4 | 591 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133323.1 | NP_579857.1 | 4 | 591 | ||
GenBank mRNA | U27186.1 | - | 4 | 591 |