siRNA Details
Gene Symbol: Map2k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031643.4 | NP_113831.1 | 9 | 1034 | ||
GenBank mRNA | BC089772.1 | - | 9 | 1028 | 7382030 | |
D13341.1 | - | 9 | 1017 | |||
D14591.1 | - | 9 | 987 | |||
X62313.1 | - | 9 | 1024 | |||
Z16415.1 | - | 9 | 1017 |
