siRNA Details
| Gene Symbol: Map2k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031643.4 | NP_113831.1 | 11 | 1181 | ||
| GenBank mRNA | BC089772.1 | - | 11 | 1175 | 7382030 | |
| D13341.1 | - | 11 | 1164 | |||
| D14591.1 | - | 11 | 1134 | |||
| X62313.1 | - | 11 | 1171 | |||
| Z16415.1 | - | 11 | 1164 | |||

Map2k1
Genomic Map