siRNA Details
| Gene Symbol: Nap1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133402.2 | NP_596893.1 | 1 | 1580 | ||
| GenBank mRNA | AB067678.1 | - | 1 | 1510 | ||
| BC101932.1 | - | 1 | 1580 | 7938895 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nap1l3| Gene Symbol: Nap1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133402.2 | NP_596893.1 | 1 | 1580 | ||
| GenBank mRNA | AB067678.1 | - | 1 | 1510 | ||
| BC101932.1 | - | 1 | 1580 | 7938895 | ||