siRNA Details
| Gene Symbol: Map4k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133407.1 | NP_596898.1 | 24 | 1887 | ||
| XM_006239656.2 | XP_006239718.1 | 25 | 2074 | |||
| GenBank mRNA | AF312224.1 | - | 24 | 1719 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map4k3| Gene Symbol: Map4k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133407.1 | NP_596898.1 | 24 | 1887 | ||
| XM_006239656.2 | XP_006239718.1 | 25 | 2074 | |||
| GenBank mRNA | AF312224.1 | - | 24 | 1719 | ||