siRNA Details
| Gene Symbol: Clcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133414.1 | NP_596905.1 | 6 | 658 | ||
| XM_006233127.1 | XP_006233189.1 | 6 | 697 | |||
| XM_006233128.2 | XP_006233190.1 | 6 | 756 | |||
| XM_008761365.1 | XP_008759587.1 | 6 | 756 | |||
| XM_008761366.1 | XP_008759588.1 | 5 | 595 | |||
| GenBank mRNA | AB052919.1 | - | 4 | 514 | ||
| AB052920.1 | - | 4 | 514 | |||
| AB052921.1 | - | 5 | 616 | |||
| AB052922.1 | - | 6 | 658 | |||
| AB052923.1 | - | 6 | 678 | |||
| AF117330.1 | - | 6 | 646 | |||
| BC081736.1 | - | 6 | 734 | 7128078 | ||

Clcc1
Genomic Map